Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sntg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172951.3 | NP_766539.2 | 9 | 808 | ||
GenBank mRNA | AF367760.1 | - | 9 | 663 | ||
AK008673.1 | - | 9 | 716 | |||
AK035310.1 | - | 9 | 762 | |||
AK087560.1 | - | 9 | 758 | |||
AK137345.1 | - | 9 | 767 | |||
BC138956.1 | - | 9 | 722 | 8861978 | ||
BC145476.1 | - | 9 | 717 |