Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145921.1 | NP_001139393.1 | 2 | 310 | ||
NM_001145922.1 | NP_001139394.1 | 2 | 310 | |||
NM_198249.4 | NP_937892.2 | 2 | 310 | |||
GenBank mRNA | AK053592.1 | - | 2 | 243 | ||
AK131136.1 | - | 2 | 279 | |||
AK134295.1 | - | 2 | 299 | |||
AK143527.1 | - | 2 | 285 | |||
AK147761.1 | - | 2 | 246 | |||
AK155659.1 | - | 2 | 178 | |||
AK163945.1 | - | 2 | 311 | |||
BC059018.1 | - | 2 | 212 | 6848691 |
Gene Symbol: Gm16617 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK136665.1 | - | 2 | 226 |