Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081280.1 | NP_001074749.1 | 12, 13 | 3051 | ||
NM_175547.3 | NP_780756.1 | 9, 10 | 974 | |||
GenBank mRNA | AK079766.1 | - | 9, 10 | 898 | ||
AK084199.1 | - | 9, 10 | 888 | |||
AK171971.1 | - | 11, 12 | 4064 | |||
BC130223.1 | - | 9, 10 | 2440 | |||
BC152543.1 | - | 9, 10 | 2449 |