Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012007.1 | NP_036137.1 | 6 | 727 | ||
GenBank mRNA | AF136272.1 | - | 6 | 727 | ||
AF158182.1 | - | 5 | 667 | |||
AK005526.1 | - | 6 | 734 | |||
AK131661.1 | - | 6 | 734 | |||
BC103769.1 | - | 6 | 723 | 30787117 |