Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Colgalt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177756.4 | NP_808424.3 | 6, 7 | 1208 | ||
GenBank mRNA | AK052838.1 | - | 6, 7 | 1209 | ||
AK076222.1 | - | 6, 7 | 950 | |||
AK085716.1 | - | 6, 7 | 1248 | |||
AK134084.1 | - | 6, 7 | 1189 | |||
BC068118.1 | - | 6, 7 | 1154 | 5706069 |