Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgat4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290801.1 | NP_001277730.1 | 4 | 690 | ||
NM_173870.3 | NP_776295.1 | 5 | 769 | |||
GenBank mRNA | AB053217.1 | - | 4 | 445 | ||
AK052278.1 | - | 2 | 266 | |||
AK155724.1 | - | 5 | 770 | |||
AK163327.1 | - | 4 | 522 |