Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175031.3 | NP_778196.2 | 17 | 2305 | ||
GenBank mRNA | AK029842.1 | - | 17 | 2215 | ||
AK047284.1 | - | 6 | 758 | |||
AK086716.1 | - | 1 | 93 | |||
AK173142.1 | - | 16 | 2171 | |||
BC043103.1 | - | 17 | 2257 | 6406485 | ||
BC058698.1 | - | 14 | 1986 | 6831702 | ||
BC061470.1 | - | 16 | 2114 |