Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aifm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290364.1 | NP_001277293.1 | 9 | 1113 | ||
NM_012019.3 | NP_036149.1 | 9 | 1125 | |||
GenBank mRNA | AF100927.1 | - | 9 | 951 | ||
AK133764.1 | - | 9 | 965 | |||
BC003292.1 | - | 9 | 988 | 3500037 | ||
BC024882.1 | - | 9 | 1609 | |||
DQ016499.1 | - | 9 | 988 |