Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r5e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012024.2 | NP_036154.1 | 8 | 1372 | ||
GenBank mRNA | AK053228.1 | - | 7 | 1223 | ||
AK088366.1 | - | 8 | 1344 | |||
AK132432.1 | - | 8 | 1405 | |||
AK133824.1 | - | 7 | 1315 | |||
AK220163.1 | - | 8 | 1362 | |||
BC019199.1 | - | 2 | 97 | |||
BC085149.1 | - | 8 | 1372 | 6848033 | ||
BC094387.1 | - | 8 | 1364 | |||
U49728.1 | - | 6 | 652 |