Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147153.3 | NP_671495.1 | 8 | 689 | ||
NM_178851.3 | NP_849182.1 | 7 | 656 | |||
GenBank mRNA | AB028844.1 | - | 7 | 636 | ||
AF281050.1 | - | 8 | 666 | |||
AF281051.1 | - | 7 | 633 | |||
AK038824.1 | - | 7 | 645 | |||
AK042253.1 | - | 7 | 645 | |||
AK084743.1 | - | 7 | 656 | |||
AK129210.1 | - | 7 | 651 | |||
AK140041.1 | - | 8 | 694 | |||
AK141424.1 | - | 7 | 639 | |||
AK154103.1 | - | 7 | 645 | |||
AK159889.1 | - | 7 | 649 | |||
AK160109.1 | - | 7 | 649 | |||
AK170831.1 | - | 8 | 716 | |||
AK171008.1 | - | 7 | 645 | |||
BC007479.1 | - | 7 | 607 | 3492875 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK156864.1 | - | 36 | Chr. - - [-] on Build |