Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Racgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253808.1 | NP_001240737.1 | 9 | 966 | ||
NM_001253809.1 | NP_001240738.1 | 9 | 962 | |||
NM_012025.7 | NP_036155.1 | 10 | 1097 | |||
GenBank mRNA | AB030252.1 | - | 9 | 946 | ||
AF079974.1 | - | 9 | 907 | |||
AF212320.1 | - | 9 | 933 | |||
AF212321.1 | - | 10 | 1064 | |||
AK053850.1 | - | 9 | 928 | |||
AK144608.1 | - | 10 | 1065 | |||
AK144745.1 | - | 9 | 926 | |||
AK154929.1 | - | 10 | 1085 | |||
AK162660.1 | - | 7 | 4166 | |||
AK165897.1 | - | 9 | 919 | |||
AK166084.1 | - | 10 | 1055 | |||
AK166094.1 | - | 9 | 924 | |||
AK168020.1 | - | 9 | 923 | |||
AK168025.1 | - | 9 | 923 | |||
AK168170.1 | - | 9 | 923 | |||
AK168679.1 | - | 9 | 907 | |||
AK180378.1 | - | 1 | 33 | |||
AK186613.1 | - | 1 | 33 | |||
BC010715.1 | - | 9 | 912 | 3602242 |