Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spag1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012031.3 | NP_036161.2 | 19 | 2926 | ||
GenBank mRNA | AF181252.1 | - | 19 | 2918 | ||
AF181253.1 | - | 9 | 1610 | |||
AK032601.1 | - | 16 | 2721 | |||
BC046313.1 | - | 19 | 2926 | 6310604 | ||
BC098201.1 | - | 3 | 2256 |