Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrig2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025067.1 | NP_001020238.1 | 1, 2 | 402 | ||
GenBank mRNA | AK036116.1 | - | 1, 2 | 403 | ||
AK155089.1 | - | 1, 2 | 465 | |||
AK158022.1 | - | 2, 3 | 419 | |||
BC094228.1 | - | 1, 2 | 339 | 30362610 | ||
BC138422.1 | - | 1, 2 | 313 | 8861442 | ||
CD355189.1 | - | 2, 3 | 257 |