Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009503.4 | NP_033529.3 | 6 | 1006 | ||
GenBank mRNA | AK028264.1 | - | 6 | 935 | ||
AK030751.1 | - | 6 | 837 | |||
AK083821.1 | - | 4 | 512 | |||
AK146109.1 | - | 6 | 903 | |||
AK149931.1 | - | 6 | 899 | |||
AK151109.1 | - | 6 | 900 | |||
AK151418.1 | - | 6 | 900 | |||
AK153012.1 | - | 6 | 936 | |||
AK153249.1 | - | 6 | 977 | |||
AK159177.1 | - | 6 | 935 | |||
AK159475.1 | - | 6 | 934 | |||
AK159509.1 | - | 6 | 978 | |||
AK167794.1 | - | 6 | 954 | |||
AK169140.1 | - | 6 | 908 | |||
AK189857.1 | - | 2 | 215 | |||
AK193877.1 | - | 2 | 215 | |||
AK193943.1 | - | 2 | 215 | |||
AK203529.1 | - | 2 | 215 | |||
AK209970.1 | - | 2 | 215 | |||
AK211047.1 | - | 2 | 215 | |||
AK211479.1 | - | 2 | 215 | |||
AK214199.1 | - | 2 | 215 | |||
AK214818.1 | - | 2 | 215 | |||
AK218438.1 | - | 2 | 215 | |||
AK218772.1 | - | 2 | 215 | |||
BC043053.1 | - | 6 | 910 | 6418662 | ||
BC049114.1 | - | 6 | 954 | 6810751 |