Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172304.3 | NP_758508.2 | 7 | 1708 | ||
GenBank mRNA | AK132870.1 | - | 7 | 1709 | ||
AK141288.1 | - | 6 | 1487 | |||
BC151176.1 | - | 6 | 1459 | 9088065 | ||
BC151177.1 | - | 6 | 1459 | 9088066 |