Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clspn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175554.4 | NP_780763.3 | 11, 12 | 2280 | ||
GenBank mRNA | AK136174.1 | - | 4, 5 | 1128 | ||
BC050848.1 | - | 11, 12 | 2235 | 6507692 |