Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epb4.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128606.1 | NP_001122078.1 | 13 | 2088 | ||
NM_001128607.1 | NP_001122079.1 | 13 | 1998 | |||
NM_183428.3 | NP_906273.3 | 16 | 3512 | |||
GenBank mRNA | AB370261.1 | - | 13 | 1930 | ||
AK133437.1 | - | 3 | 283 | |||
AK147266.1 | - | 8 | 1041 | |||
AK147644.1 | - | 8 | 1041 | |||
AK220462.1 | - | 13 | 1995 | |||
BC043337.1 | - | 2 | 135 | |||
BC068138.1 | - | 13 | 1958 | |||
BC079875.1 | - | 13 | 1917 | 6844082 | ||
CB526050.1 | - | 6 | 455 | |||
L00919.1 | - | 17 | 3587 | |||
LK021332.1 | - | 12 | 1306 | |||
LK021333.1 | - | 12 | 1306 |