Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epb4.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128606.1 | NP_001122078.1 | 16 | 2474 | ||
NM_001128607.1 | NP_001122079.1 | 16 | 2384 | |||
NM_183428.3 | NP_906273.3 | 19 | 3898 | |||
GenBank mRNA | AB370255.1 | - | 5 | 798 | ||
AB370256.1 | - | 5 | 399 | |||
AB370261.1 | - | 16 | 2316 | |||
AK133437.1 | - | 6 | 669 | |||
AK147339.1 | - | 3 | 3262 | |||
AK156891.1 | - | 3 | 209 | |||
AK220462.1 | - | 16 | 2381 | |||
BC018450.1 | - | 5 | 820 | |||
BC031511.1 | - | 5 | 820 | |||
BC043337.1 | - | 6 | 971 | |||
BC068138.1 | - | 16 | 2344 | |||
BC079875.1 | - | 16 | 2303 | 6844082 | ||
L00919.1 | - | 20 | 3973 | |||
LK021332.1 | - | 16 | 2142 | |||
LK021333.1 | - | 15 | 1692 |