Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pank4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK039866.1 | - | 1 | 145 | ||
AK050894.1 | - | 4 | 587 | |||
BC050089.1 | - | 4 | 585 | 30018732 |