Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177759.3 | NP_808427.1 | 7 | 1217 | ||
GenBank mRNA | AK082047.1 | - | 7 | 1218 | ||
BC150892.1 | - | 7 | 1007 | 9087803 | ||
BC172040.1 | - | 7 | 1005 | 9054326 |