Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177759.3 | NP_808427.1 | 12 | 1723 | ||
GenBank mRNA | AK082047.1 | - | 12 | 1724 | ||
BC060151.1 | - | 238 | ||||
BC150892.1 | - | 12 | 1513 | 9087803 | ||
BC172040.1 | - | 12 | 1511 | 9054326 |