Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mphosph9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081323.2 | NP_001074792.1 | 4 | 866 | ||
NM_001277867.1 | NP_001264796.1 | 5 | 956 | |||
GenBank mRNA | AK081178.1 | - | 4 | 715 | ||
AK084586.1 | - | 4 | 700 | |||
AK132598.1 | - | 5 | 839 | |||
AK166516.1 | - | 4 | 867 | |||
BC027168.1 | - | 3 | 625 | |||
BC076624.1 | - | 4 | 729 | |||
BC138423.1 | - | 1 | 268 | 8861443 | ||
BC145681.1 | - | 1 | 268 | 40130984 |