Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mphosph9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081323.2 | NP_001074792.1 | 17 | 2925 | ||
NM_001277867.1 | NP_001264796.1 | 18 | 3015 | |||
GenBank mRNA | BC076624.1 | - | 17 | 2788 | ||
BC138423.1 | - | 14 | 2327 | 8861443 | ||
BC145681.1 | - | 14 | 2327 | 40130984 |