Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109749.1 | NP_001103219.1 | 9 | 968 | ||
NM_001109751.1 | NP_001103221.1 | 8 | 1056 | |||
NM_173004.3 | NP_766592.2 | 9 | 968 | |||
GenBank mRNA | AK036262.1 | - | 9 | 938 | ||
AK043507.1 | - | 6 | 913 | |||
AK163520.1 | - | 6 | 761 | |||
AK173340.1 | - | 2 | 87 | |||
BC046975.1 | - | 6 | 590 | |||
BC115766.1 | - | 9 | 858 | 40060288 |