Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssc5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173008.2 | NP_766596.1 | 10 | 2306 | ||
GenBank mRNA | AK079906.1 | - | 10 | 2307 | ||
AK165666.1 | - | 10 | 2368 | |||
BC141044.1 | - | 10 | 2304 | 9055656 | ||
EU850434.1 | - | 10 | 2307 |