Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Orai3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198424.3 | NP_940816.1 | 2 | 950 | ||
GenBank mRNA | AB271216.1 | - | 2 | 776 | ||
AK037138.1 | - | 3 | 1094 | |||
AK136955.1 | - | 2 | 951 | |||
AM712358.1 | - | 2 | 776 | |||
BC061259.1 | - | 2 | 920 | 30251946 |