Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam155a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173446.2 | NP_775622.1 | 1 | 37 | ||
GenBank mRNA | AK032202.1 | - | 1 | 37 | ||
BC117535.1 | - | 1 | 35 | 40106181 |