Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mon1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048143.2 | NP_001041608.1 | 6 | 1777 | ||
NM_173015.3 | NP_766603.1 | 5 | 1967 | |||
GenBank mRNA | AK030288.1 | - | 5 | 1933 | ||
AK157390.1 | - | 6 | 1780 | |||
AK173047.1 | - | 5 | 1639 | |||
BC062639.1 | - | 5 | 1643 | |||
BQ831297.1 | - | 3 | 380 |