Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amica1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005421.4 | NP_001005421.3 | 4 | 415 | ||
GenBank mRNA | AY093688.1 | - | 4 | 415 | ||
BC050133.1 | - | 2 | 302 | 30023491 | ||
CJ139627.1 | - | 4 | 369 | |||
CJ141417.1 | - | 2 | 170 |