Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfkfb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173019.5 | NP_766607.3 | 2, 3 | 334 | ||
GenBank mRNA | AK155172.1 | - | 2, 3 | 272 | ||
AY646092.2 | - | 2, 3 | 220 | |||
AY707862.1 | - | 2, 3 | 237 | |||
AY756065.1 | - | 2, 3 | 220 | |||
AY756066.1 | - | 2, 3 | 220 | |||
AY756067.1 | - | 2, 3 | 220 | |||
BC057594.1 | - | 2, 3 | 286 | 6810667 | ||
CA316033.1 | - | 2, 3 | 286 | |||
DQ350638.1 | - | 3, 4 | 306 | |||
EU571215.1 | - | 2, 3 | 334 | |||
EU571216.1 | - | 4, 5 | 493 | |||
EU571217.1 | - | 2, 3 | 333 |