Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spin4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178753.4 | NP_848868.1 | 1 | 627 | ||
GenBank mRNA | AK036158.1 | - | 1 | 628 | ||
AK043030.1 | - | 1 | 595 | |||
AK047896.1 | - | 1 | 588 | |||
AK147852.1 | - | 1 | 594 | |||
AK155170.1 | - | 1 | 567 | |||
BC027796.1 | - | 1 | 593 | 5323183 |