Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 5031414D18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198642.2 | NP_941044.3 | 10 | 1593 | ||
GenBank mRNA | AK030303.1 | - | 5 | 2705 | ||
AK155201.1 | - | 10 | 1579 | |||
AK170426.1 | - | 10 | 1557 | |||
BC052668.1 | - | 10 | 1605 | 30054870 |