Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178755.3 | NP_848870.2 | 7, 8 | 745 | ||
GenBank mRNA | AK029744.1 | - | 7, 8 | 778 | ||
AK029932.1 | - | 7, 8 | 746 | |||
AK030138.1 | - | 7, 8 | 733 | |||
AK030182.1 | - | 7, 8 | 792 | |||
AK040259.1 | - | 7, 8 | 681 | |||
AK079691.1 | - | 7, 8 | 682 | |||
DQ867026.1 | - | 7, 8 | 778 | |||
DQ867027.1 | - | 7, 8 | 778 | |||
DQ867028.1 | - | 7, 8 | 778 | |||
DQ867029.1 | - | 7, 8 | 778 | |||
DQ867030.1 | - | 6, 7 | 724 |