Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lppr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178756.4 | NP_848871.1 | 7 | 1057 | ||
GenBank mRNA | AK034251.1 | - | 7 | 1152 | ||
AK043762.1 | - | 7 | 959 | |||
AK047614.1 | - | 7 | 987 | |||
AK087439.1 | - | 7 | 1208 | |||
AK087514.1 | - | 7 | 1058 | |||
AK160897.1 | - | 7 | 1184 | |||
AY345342.1 | - | 7 | 1057 | |||
BC096762.1 | - | 7 | 1033 | 30094664 |