Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190717.1 | NP_001177646.1 | 12 | 2047 | ||
NM_013726.3 | NP_038754.1 | 12 | 2050 | |||
GenBank mRNA | AJ003132.1 | - | 12 | 1940 | ||
BC138709.1 | - | 12 | 1868 | 8861731 | ||
BC138710.1 | - | 12 | 1868 | 8861732 | ||
BC145392.1 | - | 12 | 1865 | 9053950 |