Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Azi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048146.2 | NP_001041611.1 | 5 | 946 | ||
NM_001286507.1 | NP_001273436.1 | 4 | 846 | |||
NM_001286508.1 | NP_001273437.1 | 5 | 864 | |||
NM_013727.4 | NP_038755.1 | 5 | 946 | |||
GenBank mRNA | AB007141.1 | - | 5 | 782 | ||
AK004992.1 | - | 5 | 925 | |||
AK015942.1 | - | 5 | 767 | |||
AK030076.1 | - | 5 | 778 | |||
AK036102.1 | - | 4 | 753 | |||
AK134714.1 | - | 5 | 849 | |||
AK153477.1 | - | 5 | 910 | |||
AK165549.1 | - | 5 | 832 | |||
AK167783.1 | - | 5 | 890 | |||
BC006851.1 | - | 5 | 841 | 3156857 |