Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trp53bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290830.1 | NP_001277759.1 | 13 | 2905 | ||
NM_013735.4 | NP_038763.3 | 13 | 2905 | |||
GenBank mRNA | AJ414734.1 | - | 12 | 2766 | ||
BC079906.1 | - | 13 | 2839 | 30639375 |