Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbcel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173038.3 | NP_766626.1 | 8 | 1272 | ||
GenBank mRNA | AK052261.1 | - | 8 | 1241 | ||
AK077017.1 | - | 8 | 1274 | |||
AK084854.1 | - | 8 | 1286 | |||
AK153922.1 | - | 2 | 262 | |||
BC047462.1 | - | 8 | 1139 | |||
BC139386.1 | - | 8 | 1270 | 8862408 | ||
BC139387.1 | - | 8 | 1270 | 8862409 |