Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esyt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177775.3 | NP_808443.2 | 8, 9 | 1074 | ||
GenBank mRNA | AK086367.1 | - | 8, 9 | 1075 | ||
AK220532.1 | - | 8, 9 | 1062 | |||
BC118967.2 | - | 1022 | 40097262 |