Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nufip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013745.5 | NP_038773.1 | 3, 4 | 620 | ||
GenBank mRNA | AF159549.1 | - | 3, 4 | 578 | ||
AK009260.1 | - | 3, 4 | 596 | |||
AK032324.1 | - | 3, 4 | 621 | |||
AK156205.1 | - | 3, 4 | 607 | |||
BC020364.1 | - | 3, 4 | 595 | |||
BC056192.1 | - | 3, 4 | 606 | 30024494 |