Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nufip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013745.5 | NP_038773.1 | 9, 10 | 1384 | ||
GenBank mRNA | AF159549.1 | - | 9, 10 | 1342 | ||
AK032324.1 | - | 9, 10 | 1385 | |||
AK156205.1 | - | 9, 10 | 1371 | |||
BC020364.1 | - | 9, 10 | 1359 | |||
BC056192.1 | - | 9, 10 | 1370 | 30024494 |