Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nbn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013752.3 | NP_038780.3 | 10 | 1401 | ||
GenBank mRNA | AB016988.1 | - | 10 | 1323 | ||
AF076687.1 | - | 10 | 1395 | |||
AF092840.1 | - | 10 | 1384 | |||
AK031933.1 | - | 10 | 1387 | |||
AK132302.1 | - | 10 | 1442 | |||
AK134805.1 | - | 10 | 1437 | |||
AK134960.1 | - | 10 | 1429 | |||
AK159790.1 | - | 10 | 1408 | |||
AK160614.1 | - | 10 | 1368 | |||
BC003719.1 | - | 10 | 1410 | |||
BC013190.1 | - | 10 | 1401 | |||
BC044773.2 | - | 10 | 1400 | 5067851 | ||
BC055061.1 | - | 10 | 1393 | 6528890 |