Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gyg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013755.3 | NP_038783.1 | 3, 4 | 429 | ||
GenBank mRNA | AA553085.1 | - | 3, 4 | 428 | ||
AF114031.1 | - | 3, 4 | 364 | |||
AK159576.1 | - | 3, 4 | 410 | |||
AK201075.1 | - | 1, 2 | 147 | |||
AK208954.1 | - | 1, 2 | 147 | |||
AK212191.1 | - | 1, 2 | 147 | |||
AK214082.1 | - | 1, 2 | 147 | |||
BC029903.1 | - | 3, 4 | 390 | 4487769 |