Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yme1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013771.5 | NP_038799.1 | 17, 18 | 2121 | ||
GenBank mRNA | AF090430.1 | - | 17, 18 | 2099 | ||
AK045141.1 | - | 17, 18 | 2105 | |||
AK079175.1 | - | 17, 18 | 2081 | |||
AY136286.1 | - | 17, 18 | 1937 | |||
AY136287.1 | - | 17, 18 | 2090 | |||
BC007128.1 | - | 17, 18 | 2082 | 3591123 |