Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007268.1 | NP_001007269.1 | 6 | 1021 | ||
NM_013849.3 | NP_038877.2 | 2 | 346 | |||
GenBank mRNA | AB027003.1 | - | 1 | 127 | ||
AF237620.1 | - | 2 | 329 | |||
AK006247.1 | - | 2 | 347 | |||
AY674052.1 | - | 6 | 1021 | |||
BC100421.1 | - | 1 | 252 | 30919819 |