Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025302.3 | NP_079578.1 | 7 | 801 | ||
GenBank mRNA | AK007672.1 | - | 7 | 789 | ||
AK009521.1 | - | 7 | 795 | |||
BC027762.1 | - | 7 | 802 | 5353459 |