Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030239.2 | NP_084515.2 | 12 | 1558 | ||
GenBank mRNA | AF324242.1 | - | 12 | 1510 | ||
AK030831.1 | - | 12 | 1560 | |||
AK051880.1 | - | 12 | 1524 | |||
AK156626.1 | - | 12 | 1526 | |||
BC029095.1 | - | 10 | 1313 | |||
BC138772.1 | - | 12 | 1539 | 8861794 | ||
BC138773.1 | - | 12 | 1538 | 8861795 |