Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031884.1 | NP_114090.1 | 2 | 294 | ||
GenBank mRNA | AF312713.2 | - | 2 | 382 | ||
AK149569.1 | - | 2 | 265 | |||
AY195872.1 | - | 2 | 382 | |||
AY195873.1 | - | 2 | 382 | |||
BC106766.2 | - | 2 | 271 | 40039484 |