Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abca3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039581.2 | NP_001034670.1 | 10 | 1708 | ||
NM_013855.3 | NP_038883.2 | 11 | 1781 | |||
GenBank mRNA | AF213394.1 | - | 1 | 95 | ||
AK089316.1 | - | 10 | 1628 | |||
AK089794.1 | - | 11 | 1722 | |||
AK154698.1 | - | 11 | 1689 | |||
AK165458.1 | - | 11 | 1717 | |||
AK166355.1 | - | 10 | 1688 | |||
AK168428.1 | - | 10 | 2274 | |||
AY083616.1 | - | 11 | 1676 | |||
BC042663.1 | - | 5 | 723 | |||
BC047356.1 | - | 11 | 1746 | |||
BC079617.1 | - | 8 | 1272 | 30362309 |