Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gemin4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177367.3 | NP_796341.2 | 2 | 1289 | ||
GenBank mRNA | AK133821.1 | - | 2 | 1382 | ||
AK141078.1 | - | 2 | 1268 | |||
AK154061.1 | - | 2 | 1270 | |||
AK165864.1 | - | 2 | 1233 | |||
BC031425.1 | - | 2 | 1229 | 3982927 | ||
BC062157.1 | - | 2 | 1237 | 6508223 |
Gene Symbol: Fam57a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK030014.1 | - | 928 | |||
AK036850.1 | - | 1387 | ||||
AK158967.1 | - | 2079 |